Assay Details
| Gene Symbol: Myl12b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017343.2 | NP_059039.2 | 1 - 2 | 77 | 86 | |
| GenBank mRNA | BC060577.1 | - | 1 - 2 | 76 | 6889406 | 86 |
| CF977112.1 | - | 1 - 2 | 81 | 86 | ||
| EV769828.1 | - | 1 - 2 | 64 | 86 | ||
| FQ215510.1 | - | 1 - 2 | 63 | 86 | ||
| FQ229195.1 | - | 1 - 2 | 63 | 86 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC060577.1, NM_017343.2 transcripts.
Set Membership: |
|

Myl12b
Genomic Map