Assay Details
| Gene Symbol: Map7d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289778.1 | NP_001276707.1 | 2 - 3 | 384 | 64 | |
| GenBank mRNA | AB266744.1 | - | 2 - 3 | 384 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Map7d2
Genomic Map