Assay Details
| Gene Symbol: Rps6ka3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192004.1 | NP_001178933.1 | 19 - 20 | 2071 | 87 | |
| XM_006256923.2 | XP_006256985.1 | 18 - 19 | 2502 | 87 | ||
| XM_008773187.1 | XP_008771409.1 | 20 - 21 | 5484 | 87 | ||
| XM_008773188.1 | XP_008771410.1 | 21 - 22 | 2995 | 87 | ||
| XM_008773189.1 | XP_008771411.1 | 21 - 22 | 4519 | 87 | ||
| XM_008773190.1 | XP_008771412.1 | 19 - 20 | 5437 | 87 | ||
| XM_008773191.1 | XP_008771413.1 | 20 - 21 | 4472 | 87 | ||
| XM_008773192.1 | XP_008771414.1 | 19 - 20 | 4435 | 87 | ||
| XM_008773193.1 | XP_008771415.1 | 19 - 20 | 1799 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rps6ka3
Genomic Map