Assay Details
| Gene Symbol: Igbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031624.3 | NP_113812.1 | 4 - 5 | 765 | 93 | |
| GenBank mRNA | AF000577.2 | - | 4 - 5 | 757 | 93 | |
| BC068202.1 | - | 4 - 5 | 769 | 6921265 | 93 | |
| FQ222671.1 | - | 4 - 5 | 767 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Igbp1
Genomic Map