Assay Details
| Gene Symbol: Pgr15l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106583.1 | NP_001100053.1 | 1 - 2 | 175 | 142 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001106583.1 transcripts.
Set Membership: |
|

Pgr15l
Genomic Map