Assay Details
| Gene Symbol: Il1rapl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177935.2 | NP_808796.1 | 6 - 7 | 913 | 125 | |
| XM_008773261.1 | XP_008771483.1 | 1 - 2 | 673 | 125 | ||
| GenBank mRNA | AY216593.1 | - | 6 - 7 | 913 | 125 | |
More Information
Updated Mapping Information
Set Membership: |
|

Il1rapl1
Genomic Map