Assay Details
| Gene Symbol: Mpv17l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106072.2 | NP_001099542.1 | 1 - 2 | 252 | 93 | |
| XM_008771087.1 | XP_008769309.1 | 1 - 2 | 257 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mpv17l2
Genomic Map