Assay Details
| Gene Symbol: Rab8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053998.2 | NP_446450.2 | 1 - 2 | 127 | 86 | |
| GenBank mRNA | BC071176.1 | - | 1 - 2 | 120 | 86 | |
| BC105863.1 | - | 1 - 2 | 127 | 7457017 | 86 | |
| JN019799.1 | - | 1 - 2 | 122 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rab8a
Genomic Map