Assay Details
| Gene Symbol: Rab8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053998.2 | NP_446450.2 | 4 - 5 | 332 | 78 | |
| GenBank mRNA | BC071176.1 | - | 4 - 5 | 325 | 78 | |
| BC087584.1 | - | 3 - 4 | 163 | 78 | ||
| BC105863.1 | - | 4 - 5 | 332 | 7457017 | 78 | |
| JN019799.1 | - | 4 - 5 | 327 | 78 | ||
| M83675.1 | - | 2 - 3 | 111 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rab8a
Genomic Map