Assay Details
| Gene Symbol: Tmem38a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100175.1 | NP_001093645.1 | 4 - 5 | 789 | 67 | |
| GenBank mRNA | BC161801.1 | - | 392 | 7128088 | 67 | |
| EF690436.1 | - | 4 - 5 | 789 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmem38a
Genomic Map