Assay Details
| Gene Symbol: Tmem59l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271055.1 | NP_001257984.1 | 4 - 5 | 602 | 80 | |
| XM_006252897.2 | XP_006252959.1 | 5 - 6 | 672 | 80 | ||
| GenBank mRNA | FQ212329.1 | - | 4 - 5 | 576 | 80 | |
| FQ214027.1 | - | 4 - 5 | 556 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmem59l
Genomic Map