Assay Details
| Gene Symbol: Ocel1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106065.1 | NP_001099535.1 | 1 - 2 | 700 | 64 | |
| XM_006252856.2 | XP_006252918.1 | 1 - 2 | 1213 | 64 | ||
| XM_008771079.1 | XP_008769301.1 | 1 - 2 | 1215 | 64 | ||
| GenBank mRNA | BC169113.1 | - | 1 - 2 | 675 | 7445881 | 64 |
More Information
Updated Mapping Information
Set Membership: |
|

Ocel1
Genomic Map