Assay Details
| Gene Symbol: Exoc6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109246.1 | NP_001102716.1 | 1 - 2 | 179 | 62 | |
| XM_008763075.1 | XP_008761297.1 | 1 - 2 | 216 | 62 | ||
| GenBank mRNA | BC168850.1 | - | 1 - 2 | 146 | 8362904 | 62 |
More Information
Updated Mapping Information
Set Membership: |
|

Exoc6b
Genomic Map