Assay Details
| Gene Symbol: Dok1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025416.1 | NP_001020587.1 | 3 - 4 | 460 | 70 | |
| XM_008763022.1 | XP_008761244.1 | 3 - 4 | 502 | 70 | ||
| GenBank mRNA | BC097972.1 | - | 3 - 4 | 460 | 7442066 | 70 |
More Information
Updated Mapping Information
Set Membership: |
|

Dok1
Genomic Map