Assay Details
| Gene Symbol: NEWGENE_1562258 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008763087.1 | XP_008761309.1 | 2 - 3 | 408 | 90 | |
| XM_008763088.1 | XP_008761310.1 | 2 - 3 | 408 | 90 | ||
| XM_008763089.1 | XP_008761311.1 | 2 - 3 | 646 | 90 | ||
| Gene Symbol: Ino80b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048233.1 | NP_001041698.1 | 259 | 90 | ||
| GenBank mRNA | BC105625.1 | - | 259 | 7364756 | 90 | |
| FQ214280.1 | - | 646 | 90 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

NEWGENE_1562258
Genomic Map