Assay Details
| Gene Symbol: Cidec | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024333.2 | NP_001019504.2 | 2 - 3 | 101 | 60 | |
| NM_001244797.1 | NP_001231726.1 | 2 - 3 | 113 | 60 | ||
| NM_001244798.1 | NP_001231727.1 | 2 - 3 | 102 | 60 | ||
| GenBank mRNA | BC083860.1 | - | 2 - 3 | 101 | 7126532 | 60 |
| FM043477.1 | - | 2 - 3 | 110 | 60 | ||
| FM103335.1 | - | 2 - 3 | 102 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cidec
Genomic Map