Assay Details
| Gene Symbol: Snapc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109643.2 | NP_001103113.1 | 1 - 2 | 117 | 78 | |
| GenBank mRNA | EV775546.1 | - | 1 - 2 | 153 | 78 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Snapc5
Genomic Map