Assay Details
| Gene Symbol: Npat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108147.1 | NP_001101617.1 | 4 - 5 | 513 | 56 | |
| XM_008766264.1 | XP_008764486.1 | 2 - 3 | 1234 | 56 | ||
| XM_008766265.1 | XP_008764487.1 | 2 - 3 | 1234 | 56 | ||
| XM_008766266.1 | XP_008764488.1 | 2 - 3 | 1234 | 56 | ||
| XM_008766267.1 | XP_008764489.1 | 2 - 3 | 1234 | 56 | ||
| XM_008766268.1 | XP_008764490.1 | 4 - 5 | 402 | 56 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Npat
Genomic Map