Assay Details
| Gene Symbol: Snhg11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258011.1 | NP_001244940.1 | 3 - 4 | 533 | 106 | |
| GenBank mRNA | BC094214.1 | - | 3 - 4 | 533 | 7314456 | 106 |
More Information
Updated Mapping Information
Set Membership: |
|

Snhg11
Genomic Map