Assay Details
| Gene Symbol: Pcif1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108605.1 | NP_001102075.1 | 10 - 11 | 2516 | 83 | |
| XM_008762514.1 | XP_008760736.1 | 9 - 10 | 1133 | 83 | ||
| GenBank mRNA | FQ225793.1 | - | 10 - 11 | 1312 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pcif1
Genomic Map