Assay Details
| Gene Symbol: Oser1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201560.2 | NP_963854.1 | 1 - 2 | 77 | 123 | |
| GenBank mRNA | AJ621831.1 | - | 1 - 2 | 81 | 123 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AJ621831.1,NM_201560.2 transcripts.
Set Membership: |
|

Oser1
Genomic Map