Assay Details
| Gene Symbol: RGD1564534 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028321.1 | 1 | 384 | 63 | ||
| GenBank mRNA | BC111405.1 | - | 1 | 384 | 7446493 | 63 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

RGD1564534
Genomic Map