Assay Details
| Gene Symbol: Fam19a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134700.1 | NP_001128172.1 | 2 - 3 | 347 | 98 | |
| GenBank mRNA | BC160856.1 | - | 2 - 3 | 347 | 8364977 | 98 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC160856.1, NM_001134700.1 transcripts.
Set Membership: |
|

Fam19a4
Genomic Map