Assay Details
| Gene Symbol: Slc41a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037492.1 | NP_001032569.1 | 5 - 6 | 884 | 60 | |
| XM_006236871.2 | XP_006236933.1 | 6 - 7 | 894 | 60 | ||
| XM_006236872.2 | XP_006236934.1 | 6 - 7 | 951 | 60 | ||
| XM_006236873.1 | XP_006236935.1 | 6 - 7 | 1268 | 60 | ||
| XM_006236874.2 | XP_006236936.1 | 5 - 6 | 823 | 60 | ||
| XM_006236875.2 | XP_006236937.1 | 6 - 7 | 1100 | 60 | ||
| XM_006236876.2 | XP_006236938.1 | 5 - 6 | 495 | 60 | ||
| XM_006236877.2 | XP_006236939.1 | 5 - 6 | 1240 | 60 | ||
| XM_006236878.2 | XP_006236940.1 | 5 - 6 | 541 | 60 | ||
| XM_006236879.2 | XP_006236941.1 | 5 - 6 | 536 | 60 | ||
| XM_008763134.1 | XP_008761356.1 | 6 - 7 | 1095 | 60 | ||
| GenBank mRNA | BC104698.1 | - | 5 - 6 | 884 | 7134770 | 60 |
More Information
Updated Mapping Information
Set Membership: |
|

Slc41a3
Genomic Map