Assay Details
| Gene Symbol: Cog2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110494.1 | NP_001103964.1 | 2 - 3 | 306 | 58 | |
| XM_006255816.2 | XP_006255878.1 | 2 - 3 | 324 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cog2
Genomic Map