Assay Details
| Gene Symbol: Fa2h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135583.1 | NP_001129055.1 | 1 - 2 | 340 | 63 | |
| GenBank mRNA | DQ339484.1 | - | 1 - 2 | 340 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
sebaceous gland cell differentiation
fatty acid metabolic process
fatty acid biosynthetic process
sphingolipid metabolic process
lipid modification
central nervous system myelin maintenance
peripheral nervous system myelin maintenance
regulation of cell proliferation
regulation of hair cycle
oxidation-reduction process
iron ion binding
heme binding
fatty acid alpha-hydroxylase activity
sebaceous gland cell differentiation
fatty acid metabolic process
fatty acid biosynthetic process
sphingolipid metabolic process
lipid modification
central nervous system myelin maintenance
peripheral nervous system myelin maintenance
regulation of cell proliferation
regulation of hair cycle
oxidation-reduction process
iron ion binding
heme binding
fatty acid alpha-hydroxylase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P712297 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P16988 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 898777 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 37937 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 263095 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 291645 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 635532 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 762389 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 468622 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 35191 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 729950 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 117101 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 390836 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 699566 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 743561 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535880 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 536714 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 257026 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 862646 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 261887 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 826450 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 129946 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427154 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308825 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 544560 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 290669 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 859015 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488722 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504071 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 267694 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 92578 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 465835 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 403640 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 161635 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 788582 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 386089 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 592087 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 113674 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 88534 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 69645 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 82821 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140652 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 2797 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P16988 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P712297 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P712297 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P16988 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P712297 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P16988 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P16988 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 15307 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10811132 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10811127 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10811133 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10811130 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10811134 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10811131 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10811129 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10811128 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10811126 | - | - |
| Rat Genome 230 2.0 Array | 1393808_at | - | - |
| Rat Genome 230 2.0 Array | 1393808_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1393808_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1393808_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1900012066.rn.1 | TC1900001471.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1900012067.rn.1 | TC1900001471.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1900012068.rn.1 | TC1900001471.rn.1 | RTA TC |
| Rat Expression Set 230 | 1393808_at | - | - |
| Rat Expression Set 230 | 1393808_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10811126 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21068989 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1353545 | - | - |

Fa2h
Genomic Map