Assay Details
| Gene Symbol: Nfatc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108447.1 | NP_001101917.1 | 8 - 9 | 2187 | 87 | |
| XM_006255513.2 | XP_006255575.1 | 8 - 9 | 2373 | 87 | ||
| XM_008772518.1 | XP_008770740.1 | 8 - 9 | 2374 | 87 | ||
| XM_008772519.1 | XP_008770741.1 | 8 - 9 | 2373 | 87 | ||
| XM_008772520.1 | XP_008770742.1 | 8 - 9 | 2373 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
patterning of blood vessels
response to hypoxia
blood vessel remodeling
regulation of transcription, DNA-templated
heart development
cardiac muscle hypertrophy in response to stress
myotube differentiation
myotube cell development
T cell differentiation
calcineurin-NFAT signaling cascade
cellular respiration
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
thymus development
skeletal muscle fiber development
smooth muscle cell differentiation
muscle cell development
cellular response to calcium ion
cellular response to lithium ion
regulation of store-operated calcium entry
RNA polymerase II core promoter proximal region sequence-specific DNA binding
core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription factor binding
sequence-specific DNA binding
patterning of blood vessels
response to hypoxia
blood vessel remodeling
regulation of transcription, DNA-templated
heart development
cardiac muscle hypertrophy in response to stress
myotube differentiation
myotube cell development
T cell differentiation
calcineurin-NFAT signaling cascade
cellular respiration
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
thymus development
skeletal muscle fiber development
smooth muscle cell differentiation
muscle cell development
cellular response to calcium ion
cellular response to lithium ion
regulation of store-operated calcium entry
RNA polymerase II core promoter proximal region sequence-specific DNA binding
core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription factor binding
sequence-specific DNA binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P077002 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P11151 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 508461 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 864457 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 223930 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 706932 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112843 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 74117 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 508142 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 316224 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 70487 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 314285 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 582334 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 578467 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 167231 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 462516 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907934 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 792239 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 596579 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73921 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 643224 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 165762 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586462 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 299440 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 874139 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116851 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 750277 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 159001 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 395701 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 921501 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865249 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 22493 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 901573 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 933582 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 339529 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 386450 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 514330 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 623636 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 5317 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535313 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 437428 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 684964 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 212019 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 801156 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 284237 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 111201 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 379868 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 254421 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691788 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 191460 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 157308 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 364731 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 45197 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144576 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 58195 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 38046 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 155198 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 79701 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 149411 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 157576 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 67991 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 453 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 125096 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P11151 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P077002 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P077002 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P11151 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P11151 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P077002 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P615549 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P192275 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 96141 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 33858 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10807461 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10807453 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10807460 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10807459 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10807455 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10807458 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10807463 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10807454 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10807456 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10807462 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10807457 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10807452 | - | - |
| Rat Genome 230 2.0 Array | 1374064_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1374064_at | - | - |
| Affymetrix HT Rat Focus Array | 1374064_at | - | - |
| Affymetrix HT Rat Focus Array | 1374064_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1900004346.rn.1 | TC1900000503.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1900004345.rn.1 | TC1900000503.rn.1 | RTA TC |
| Rat Expression Set 230 | 1374064_PM_at | - | - |
| Rat Expression Set 230 | 1374064_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10807452 | - | - |
| Rat Genome U34 Set | rc_AI070175_at | - | - |
| Rat Genome U34 Set | rc_AA925516_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21554692 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1353054 | - | - |

Nfatc3
Genomic Map