Assay Details
| Gene Symbol: Otud4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191700.1 | NP_001178629.1 | 13 - 14 | 1477 | 61 | |
| XM_006255402.2 | XP_006255464.1 | 13 - 14 | 1816 | 61 | ||
| XM_008772493.1 | XP_008770715.1 | 13 - 14 | 1992 | 61 | ||
| XM_008772494.1 | XP_008770716.1 | 13 - 14 | 1548 | 61 | ||
| XM_008772495.1 | XP_008770717.1 | 14 - 15 | 1596 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Otud4
Genomic Map