Assay Details
| Gene Symbol: NEWGENE_1311658 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006245524.2 | XP_006245586.1 | 165 | 95 | ||
| XM_008767395.1 | XP_008765617.1 | 165 | 95 | |||
| XM_008767396.1 | XP_008765618.1 | 165 | 95 | |||
| Gene Symbol: Pdcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106927.1 | NP_001100397.1 | 1 - 2 | 137 | 95 | |
| XM_006245571.2 | XP_006245633.1 | 1 - 2 | 165 | 95 | ||
| XM_008767366.1 | XP_008765588.1 | 1 - 2 | 165 | 95 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

NEWGENE_1311658
Genomic Map