Assay Details
| Gene Symbol: Dtymk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106925.1 | NP_001100395.1 | 2 - 3 | 462 | 70 | |
| XM_006245519.2 | XP_006245581.1 | 2 - 3 | 391 | 70 | ||
| XM_008767363.1 | XP_008765585.1 | 2 - 3 | 391 | 70 | ||
| GenBank mRNA | BC159428.1 | - | 2 - 3 | 326 | 9088913 | 70 |
| FQ225176.1 | - | 2 - 3 | 325 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dtymk
Genomic Map