Assay Details
| Gene Symbol: Slc5a4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008772932.1 | XP_008771154.1 | 3 - 4 | 313 | 65 | |
| XM_008772933.1 | XP_008771155.1 | 2 - 3 | 175 | 65 | ||
More Information
Set Membership: |
|

Slc5a4b
Genomic Map