Assay Details
| Gene Symbol: Nudt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024243.1 | NP_001019414.1 | 4 - 5 | 540 | 73 | |
| GenBank mRNA | BC093618.1 | - | 4 - 5 | 540 | 7311727 | 73 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
diphosphoinositol polyphosphate catabolic process
magnesium ion binding
diphosphoinositol-polyphosphate diphosphatase activity
inositol diphosphate tetrakisphosphate diphosphatase activity
inositol-1-diphosphate-2,3,4,5,6-pentakisphosphate diphosphatase activity
inositol-3-diphosphate-1,2,4,5,6-pentakisphosphate diphosphatase activity
inositol-5-diphosphate-1,2,3,4,6-pentakisphosphate diphosphatase activity
inositol-1,5-bisdiphosphate-2,3,4,6-tetrakisphosphate 1-diphosphatase activity
inositol-1,5-bisdiphosphate-2,3,4,6-tetrakisphosphate 5-diphosphatase activity
inositol-3,5-bisdiphosphate-2,3,4,6-tetrakisphosphate 5-diphosphatase activity
diphosphoinositol polyphosphate catabolic process
magnesium ion binding
diphosphoinositol-polyphosphate diphosphatase activity
inositol diphosphate tetrakisphosphate diphosphatase activity
inositol-1-diphosphate-2,3,4,5,6-pentakisphosphate diphosphatase activity
inositol-3-diphosphate-1,2,4,5,6-pentakisphosphate diphosphatase activity
inositol-5-diphosphate-1,2,3,4,6-pentakisphosphate diphosphatase activity
inositol-1,5-bisdiphosphate-2,3,4,6-tetrakisphosphate 1-diphosphatase activity
inositol-1,5-bisdiphosphate-2,3,4,6-tetrakisphosphate 5-diphosphatase activity
inositol-3,5-bisdiphosphate-2,3,4,6-tetrakisphosphate 5-diphosphatase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P054137 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 336231 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450905 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 248570 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 316435 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 241169 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 496099 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741841 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 140339 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 295745 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 452006 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250017 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391967 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 221928 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 303737 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 197889 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 480033 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 66999 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 206971 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255279 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 45930 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 121806 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 624132 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 774864 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 651852 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 932954 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 344749 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 878801 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 655501 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 323661 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872958 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 254209 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 378012 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 156543 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 843412 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 11507 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 331701 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 587460 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 739570 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 96733 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 166500 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 959680 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 276703 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 204753 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 636909 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 373441 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 25423 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 92717 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 656341 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 142413 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 125822 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 67920 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 124905 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 23674 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156551 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 168443 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 42688 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 60925 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 101094 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P054137 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P054137 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P054137 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P100740 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 48980 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 41433 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 93494 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 79191 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 12657 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10831869 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10831871 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10831872 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10831870 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10831873 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10831868 | - | - |
| Rat Genome 230 2.0 Array | 1372074_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1383957_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1372074_at | - | - |
| Rat Genome 230 2.0 Array | 1383957_at | - | - |
| Affymetrix HT Rat Focus Array | 1372074_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1383957_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1372074_at | - | - |
| Affymetrix HT Rat Focus Array | 1383957_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR2000010035.rn.1 | TC2000001197.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR2000010037.rn.1 | TC2000001197.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR2000010036.rn.1 | TC2000001197.rn.1 | RTA TC |
| Rat Expression Set 230 | 1372074_PM_at | - | - |
| Rat Expression Set 230 | 1383957_at | - | - |
| Rat Expression Set 230 | 1383957_PM_at | - | - |
| Rat Expression Set 230 | 1372074_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10831868 | - | - |
| Rat Genome U34 Set | rc_AA998124_f_at | - | - |
| Rat Genome U34 Set | rc_AA998124_i_at | - | - |
| Rat Genome U34 Set | rc_AA998124_r_at | - | - |
| Rat Genome U34 Set | rc_AA946349_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20970334 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1352082 | - | - |

Nudt3
Genomic Map