Assay Details
| Gene Symbol: Tff3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013042.2 | NP_037174.2 | 1 - 2 | 116 | 85 | |
| GenBank mRNA | BC078873.1 | - | 1 - 2 | 96 | 7127312 | 85 |
| BP490107.1 | - | 1 - 2 | 116 | 85 | ||
| U48825.1 | - | 1 - 2 | 89 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tff3
Genomic Map