Assay Details
| Gene Symbol: Smarcb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025728.1 | NP_001020899.1 | 5 - 6 | 799 | 88 | |
| XM_006256318.2 | XP_006256380.1 | 5 - 6 | 1585 | 88 | ||
| XM_006256319.2 | XP_006256381.1 | 4 - 5 | 877 | 88 | ||
| GenBank mRNA | BC099195.1 | - | 5 - 6 | 799 | 7383650 | 88 |
| Gene Symbol: LOC103694876 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008773843.1 | XP_008772065.1 | 5 - 6 | 1585 | 88 | |
| XM_008773844.1 | XP_008772066.1 | 5 - 6 | 1558 | 88 | ||
| XM_008773845.1 | XP_008772067.1 | 4 - 5 | 877 | 88 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Smarcb1
Genomic Map