Assay Details
| Gene Symbol: Wdr46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212491.2 | NP_997656.1 | 6 - 7 | 674 | 94 | |
| GenBank mRNA | BC085934.1 | - | 6 - 7 | 674 | 7112671 | 94 |
More Information
Updated Mapping Information
Set Membership: |
|

Wdr46
Genomic Map