Assay Details
| Gene Symbol: Phyhipl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012076.1 | NP_001012076.1 | 2 - 3 | 340 | 152 | |
| XM_006256347.2 | XP_006256409.1 | 2 - 3 | 461 | 152 | ||
| GenBank mRNA | BC078977.1 | - | 2 - 3 | 340 | 7111966 | 152 |
More Information
Updated Mapping Information
Set Membership: |
|

Phyhipl
Genomic Map