Assay Details
| Gene Symbol: Ndufv3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101011.2 | NP_001094481.1 | 2 - 3 | 265 | 153 | |
| GenBank mRNA | DV725422.1 | - | 2 - 3 | 263 | 153 | |
| FQ216920.1 | - | 2 - 3 | 213 | 153 | ||
| FQ217920.1 | - | 2 - 3 | 213 | 153 | ||
| FQ218041.1 | - | 2 - 3 | 213 | 153 | ||
| FQ223657.1 | - | 2 - 3 | 213 | 153 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ndufv3
Genomic Map