Assay Details
| Gene Symbol: LOC103694870 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008773819.1 | XP_008772041.1 | 2 - 3 | 199 | 97 | |
| Gene Symbol: LOC103690052 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008772964.1 | XP_008771186.1 | 199 | 97 | ||
| Gene Symbol: Slc5a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106383.1 | NP_001099853.1 | 7 - 8 | 755 | 97 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC103694870
Genomic Map