Assay Details
| Gene Symbol: Arid5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107624.1 | NP_001101094.1 | 744 | 54 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
kidney development
nitrogen compound metabolic process
male gonad development
female gonad development
post-embryonic development
fibroblast migration
adrenal gland development
multicellular organism growth
fat cell differentiation
platelet-derived growth factor receptor signaling pathway
cell development
muscle organ morphogenesis
skeletal system morphogenesis
positive regulation of sequence-specific DNA binding transcription factor activity
palate development
face morphogenesis
adipose tissue development
fat pad development
RNA polymerase II regulatory region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
transcription coactivator activity
transcription regulatory region DNA binding
negative regulation of transcription from RNA polymerase II promoter
kidney development
nitrogen compound metabolic process
male gonad development
female gonad development
post-embryonic development
fibroblast migration
adrenal gland development
multicellular organism growth
fat cell differentiation
platelet-derived growth factor receptor signaling pathway
cell development
muscle organ morphogenesis
skeletal system morphogenesis
positive regulation of sequence-specific DNA binding transcription factor activity
palate development
face morphogenesis
adipose tissue development
fat pad development
RNA polymerase II regulatory region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
transcription coactivator activity
transcription regulatory region DNA binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P045873 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 172946 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 114699 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 572523 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205399 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 105796 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 855317 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 221774 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 396416 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448069 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 91563 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 151726 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 18631 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337589 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 415698 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 438640 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 72961 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 909097 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 728502 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 386494 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 803325 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 452301 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560623 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 294147 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 421222 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 819119 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 506558 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 188030 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 972773 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741639 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 771816 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 340554 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 57978 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 714719 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 339001 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 944405 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 65931 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 592742 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 64296 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 363482 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 712332 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527637 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 70183 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 484465 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 495968 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 921868 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660230 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 930249 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 280447 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 38960 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 902579 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 767996 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 432165 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881308 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 140400 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 21824 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 682996 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736776 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 738584 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 726491 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 214966 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 218939 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61095 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 442204 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 478188 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741416 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 511363 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 537602 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 493119 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 331014 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 675591 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 306273 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 320424 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 35857 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 21351 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 634793 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 65393 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 356784 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 464060 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 972939 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 130527 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 433951 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 826276 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 625712 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255629 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 352403 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 87122 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 51547 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 525153 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 55001 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 603243 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 318486 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 972422 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 243361 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 628016 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 698371 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 71305 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 416606 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 449484 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 970811 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 556327 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 315231 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 515804 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 575644 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 603510 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 315201 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 287267 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565234 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 776822 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 823014 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144623 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 312476 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450257 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 898965 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61742 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873889 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 136216 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 315054 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 830061 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 184936 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 570174 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 607970 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 446592 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 636615 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 188357 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236731 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 625434 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 743000 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 584595 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667567 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 497416 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 544200 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 559671 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 167146 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 835282 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 455072 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 838948 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 643901 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 66299 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 781674 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 841630 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 556789 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 291103 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 952563 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 74188 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 61347 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 52421 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 105764 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 180065 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 11732 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 147315 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 122551 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 157821 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 23292 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 129693 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 102495 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 99666 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 135581 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 160865 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 165304 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 92104 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 94789 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 155269 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 139210 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 37339 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 41339 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46714 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 114248 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P045873 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P045873 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P045873 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P253196 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 27828 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 49613 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 11249 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 63703 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 38070 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10829781 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10829785 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10829786 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10829787 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10829789 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10829782 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10829783 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10829784 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10829790 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10829788 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10829780 | - | - |
| Rat Genome 230 2.0 Array | 1382312_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1372964_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1382312_at | - | - |
| Rat Genome 230 2.0 Array | 1372964_at | - | - |
| Affymetrix HT Rat Focus Array | 1382312_at | - | - |
| Affymetrix HT Rat Focus Array | 1382312_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR2000005134.rn.1 | TC2000000564.rn.1 | RTA TC |
| Rat Expression Set 230 | 1382312_at | - | - |
| Rat Expression Set 230 | 1382312_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10829780 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22425577 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1353523 | - | - |

Arid5b
Genomic Map