Assay Details
| Gene Symbol: Vrk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012194.1 | NP_001012194.1 | 10 - 11 | 1178 | 82 | |
| XM_006240530.2 | XP_006240592.1 | 10 - 11 | 1004 | 82 | ||
| XM_006240531.2 | XP_006240593.1 | 10 - 11 | 1034 | 82 | ||
| XM_006240532.2 | XP_006240594.1 | 10 - 11 | 1426 | 82 | ||
| XM_006240533.2 | XP_006240595.1 | 10 - 11 | 913 | 82 | ||
| XM_008764822.1 | XP_008763044.1 | 10 - 11 | 1426 | 82 | ||
| GenBank mRNA | BC079130.1 | - | 10 - 11 | 1178 | 7127355 | 82 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
endocytosis
regulation of cell shape
Wnt signaling pathway
peptidyl-serine phosphorylation
histone H3-S10 phosphorylation
protein autophosphorylation
histone H3-T3 phosphorylation
Golgi disassembly
protein kinase activity
protein serine/threonine kinase activity
ATP binding
nucleosomal histone binding
histone kinase activity (H3-S10 specific)
histone kinase activity (H3-T3 specific)
protein phosphorylation
endocytosis
regulation of cell shape
Wnt signaling pathway
peptidyl-serine phosphorylation
histone H3-S10 phosphorylation
protein autophosphorylation
histone H3-T3 phosphorylation
Golgi disassembly
protein kinase activity
protein serine/threonine kinase activity
ATP binding
nucleosomal histone binding
histone kinase activity (H3-S10 specific)
histone kinase activity (H3-T3 specific)
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P1004150 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 446303 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 524574 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 629325 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 522078 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 919055 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 514845 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83210 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168355 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 137628 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 874136 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 808139 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 732029 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 928260 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871190 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853792 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 22705 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 640225 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 173281 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 830449 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 4210 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 799933 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 417675 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619903 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 191626 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 822258 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 758942 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 915677 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731476 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 782979 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 194249 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 508919 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 923059 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 625641 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 21746 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 424397 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 477040 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 674817 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 316650 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 717448 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 329895 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 580774 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 273471 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 410288 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 133975 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 685134 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 522260 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 10791 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140031 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 178873 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 3935 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 59100 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 80548 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 31613 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 54939 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1004150 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1004150 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P1004150 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1004150 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 16656 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 64052 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 81023 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 96744 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 67444 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 36877 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10886699 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10886698 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10886708 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10886707 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10886706 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10886702 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10886705 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10886710 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10886704 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10886713 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10886703 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10886701 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10886697 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10886709 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10886700 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10886693 | - | - |
| Rat Genome 230 2.0 Array | 1392168_at | - | - |
| Rat Genome 230 2.0 Array | 1392168_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1380009_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1380009_at | - | - |
| Affymetrix HT Rat Focus Array | 1392168_at | - | - |
| Affymetrix HT Rat Focus Array | 1380009_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1392168_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1380009_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0600010077.rn.1 | TC0600001440.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600010071.rn.1 | TC0600001440.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600010074.rn.1 | TC0600001440.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600010070.rn.1 | TC0600001440.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600010078.rn.1 | TC0600001440.rn.1 | RTA TC |
| Rat Expression Set 230 | 1380009_at | - | - |
| Rat Expression Set 230 | 1380009_PM_at | - | - |
| Rat Expression Set 230 | 1392168_PM_at | - | - |
| Rat Expression Set 230 | 1392168_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10886693 | - | - |
| Rat Genome U34 Set | rc_AA924442_at | - | - |
| Rat Genome U34 Set | rc_AI072422_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21267601 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1375587 | - | - |
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1376349 | - | - |

Vrk1
Genomic Map