Assay Details
| Gene Symbol: Zfp827 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006255413.2 | XP_006255475.1 | 10 - 11 | 3276 | 85 | |
| XM_008772544.1 | XP_008770766.1 | 10 - 11 | 3294 | 85 | ||
| XM_008772545.1 | XP_008770767.1 | 10 - 11 | 3294 | 85 | ||
| XM_008772546.1 | XP_008770768.1 | 10 - 11 | 3294 | 85 | ||
| XM_008772547.1 | XP_008770769.1 | 10 - 11 | 3294 | 85 | ||
More Information
Set Membership: |
|

Zfp827
Genomic Map