Assay Details
| Gene Symbol: Gcsh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133598.2 | NP_598282.2 | 2 - 3 | 310 | 64 | |
| GenBank mRNA | BC088114.1 | - | 2 - 3 | 247 | 7321648 | 64 |
| CB609750.1 | - | 2 - 3 | 336 | 64 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gcsh
Genomic Map