Assay Details
| Gene Symbol: Slc38a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192009.1 | NP_001178938.1 | 1 - 2 | 185 | 79 | |
| XM_006255721.2 | XP_006255783.1 | 1 - 2 | 1895 | 79 | ||
| XM_006255722.2 | XP_006255784.1 | 2 - 3 | 1543 | 79 | ||
| XM_008772644.1 | XP_008770866.1 | 1 - 2 | 1895 | 79 | ||
| XM_008772645.1 | XP_008770867.1 | 1 - 2 | 1895 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc38a8
Genomic Map