Assay Details
| Gene Symbol: Lyl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007677.1 | NP_001007678.1 | 3 - 4 | 642 | 55 | |
| GenBank mRNA | BC081864.1 | - | 3 - 4 | 642 | 7096152 | 55 |
More Information
Updated Mapping Information
Set Membership: |
|

Lyl1
Genomic Map