Assay Details
| Gene Symbol: Wdr83os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006255237.1 | XP_006255299.1 | 2 - 3 | 346 | 128 | |
| GenBank mRNA | BC166543.1 | - | 2 - 3 | 222 | 7300989 | 128 |
| FQ225008.1 | - | 2 - 3 | 351 | 128 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Wdr83os
Genomic Map