Assay Details
| Gene Symbol: Agrp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033650.1 | NP_387499.1 | 4 - 5 | 414 | 67 | |
| XM_008772474.1 | XP_008770696.1 | 3 - 4 | 1115 | 67 | ||
| GenBank mRNA | AF206017.1 | - | 2 - 3 | 98 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Agrp
Genomic Map