Assay Details
| Gene Symbol: Fhod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191600.1 | NP_001178529.1 | 1 - 2 | 227 | 87 | |
| XM_008772483.1 | XP_008770705.1 | 1 - 2 | 1561 | 87 | ||
| XM_008772484.1 | XP_008770706.1 | 1 - 2 | 1561 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fhod1
Genomic Map