Assay Details
| Gene Symbol: Slc7a6os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139328.2 | NP_647544.2 | 1 - 2 | 202 | 69 | |
| GenBank mRNA | DY567141.1 | - | 1 - 2 | 202 | 69 | |
| FQ211988.1 | - | 1 - 2 | 209 | 69 | ||
| FQ220945.1 | - | 1 - 2 | 213 | 69 | ||
| FQ233662.1 | - | 1 - 2 | 211 | 69 | ||
| FQ234848.1 | - | 1 - 2 | 209 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc7a6os
Genomic Map