Assay Details
| Gene Symbol: Hsd17b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024391.1 | NP_077367.1 | 4 - 5 | 853 | 91 | |
| GenBank mRNA | BC088134.1 | - | 4 - 5 | 844 | 7326035 | 91 |
| X91234.1 | - | 4 - 5 | 853 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
placenta development
androgen biosynthetic process
estrogen biosynthetic process
response to retinoic acid
oxidation-reduction process
bone development
cellular response to metal ion
estradiol 17-beta-dehydrogenase activity
testosterone dehydrogenase (NAD+) activity
in utero embryonic development
placenta development
androgen biosynthetic process
estrogen biosynthetic process
response to retinoic acid
oxidation-reduction process
bone development
cellular response to metal ion
estradiol 17-beta-dehydrogenase activity
testosterone dehydrogenase (NAD+) activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P374638 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 35399 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 750700 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 479645 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 358383 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 683394 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 821727 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 473969 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 775935 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 652682 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 446413 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 645256 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828215 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 40447 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 841476 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 869413 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 214834 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 894910 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 609586 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 165415 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 270272 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 968153 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 572977 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887604 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 228379 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 447400 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 394541 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 217449 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 493723 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 942647 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 376011 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 201665 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 49041 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 686330 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 148902 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 829217 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 835581 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 806642 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 475812 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887442 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 393512 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 628904 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 587450 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 880610 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 676214 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 101024 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 91566 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 130112 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 145669 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 311 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 40566 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 25759 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 174659 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P374638 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P374638 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P374638 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P374638 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700606955 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 6536 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 35927 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 92262 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 17506 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 76589 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 74477 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 36718 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 700606955 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10811345 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10811344 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10811342 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10811343 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10811346 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10811341 | - | - |
| Rat Genome 230 2.0 Array | 1387156_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1387156_at | - | - |
| Affymetrix HT Rat Focus Array | 1387156_at | - | - |
| Affymetrix HT Rat Focus Array | 1387156_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1900005780.rn.1 | TC1900000696.rn.1 | RTA TC |
| Rat Expression Set 230 | 1387156_at | - | - |
| Rat Expression Set 230 | 1387156_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10811341 | - | - |
| Rat Genome U34 Set | X91234_at | - | - |
| Rat Genome U34 Set | rc_AA899367_f_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21539963 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1353855 | - | - |
| RatRef-12 Expression BeadChip | ILMN_51207 | - | - |

Hsd17b2
Genomic Map