Assay Details
| Gene Symbol: Clgn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109472.1 | NP_001102942.1 | 8 - 9 | 1105 | 91 | |
| XM_006255298.2 | XP_006255360.1 | 8 - 9 | 1058 | 91 | ||
| XM_006255299.2 | XP_006255361.1 | 8 - 9 | 1139 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Clgn
Genomic Map